Assay Details
| Gene Symbol: HCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082707.1 | NP_001076176.1 | 1332 | 63 | ||
| GenBank mRNA | AB022927.2 | - | 1332 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCN4| Gene Symbol: HCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082707.1 | NP_001076176.1 | 1332 | 63 | ||
| GenBank mRNA | AB022927.2 | - | 1332 | 63 | ||
Set Membership: |
|