Assay Details
| Gene Symbol: HMOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082747.1 | NP_001076216.1 | 769 | 129 | ||
| GenBank mRNA | S61699.1 | - | 769 | 129 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMOX2| Gene Symbol: HMOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082747.1 | NP_001076216.1 | 769 | 129 | ||
| GenBank mRNA | S61699.1 | - | 769 | 129 | ||
Set Membership: |
|