Assay Details
| Gene Symbol: ATP1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082780.1 | NP_001076249.1 | 2 - 3 | 242 | 103 | |
| GenBank mRNA | AY069937.1 | - | 2 - 3 | 242 | 103 | |
More Information
Set Membership: |
|

ATP1B2
Genomic Map