Assay Details
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082784.1 | NP_001076253.1 | 4 - 5 | 638 | 66 | |
| XM_017345498.1 | XP_017200987.1 | 5 - 6 | 720 | 66 | ||
| GenBank mRNA | L10392.1 | - | 4 - 5 | 638 | 66 | |
More Information
Set Membership: |
|

FMO4
Genomic Map