Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099969.1 | NP_001093439.1 | 671 | 95 | ||
| GenBank mRNA | AB261160.1 | - | 671 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM38A| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099969.1 | NP_001093439.1 | 671 | 95 | ||
| GenBank mRNA | AB261160.1 | - | 671 | 95 | ||
Set Membership: |
|