Assay Details
| Gene Symbol: MYH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109816.1 | NP_001103286.1 | 21 - 22 | 2994 | 68 | |
| GenBank mRNA | U32574.1 | - | 21 - 22 | 2994 | 68 | |
| Gene Symbol: LOC100303760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718960.2 | XP_002719006.1 | 22 - 23 | 2995 | 68 | |
More Information
Set Membership: |
|

MYH4
Genomic Map