Assay Details
| Gene Symbol: EDNRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105674.1 | NP_001099144.1 | 1 | 52 | 132 | |
| XM_008267274.2 | XP_008265496.1 | 2 | 535 | 132 | ||
| GenBank mRNA | AB303551.1 | - | 1 | 52 | 132 | |
More Information
Set Membership: |
|

EDNRA
Genomic Map