Assay Details
| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082131.1 | 751 | 100 | |||
| GenBank mRNA | AJ311162.1 | - | 751 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSBP1| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082131.1 | 751 | 100 | |||
| GenBank mRNA | AJ311162.1 | - | 751 | 100 | ||
Set Membership: |
|