Assay Details
Gene Symbol: CD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082177.1 | NP_001075646.1 | 1852 | 89 | ||
XM_017350537.1 | XP_017206026.1 | 2054 | 89 | |||
XM_017350538.1 | XP_017206027.1 | 1848 | 89 | |||
XM_017350539.1 | XP_017206028.1 | 1826 | 89 | |||
GenBank mRNA | AY682087.1 | - | 1852 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
