Assay Details
| Gene Symbol: CCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082294.1 | NP_001075763.1 | 1 | 44 | 69 | |
| GenBank mRNA | M57440.1 | - | 1 | 44 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082294.1 transcripts.
Set Membership: |
|

CCL2
Genomic Map