Assay Details
| Gene Symbol: INS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340618.1 | XP_017196107.1 | 52 | 87 | ||
| GenBank mRNA | U03610.1 | - | 52 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017340618.1 transcripts.
Set Membership: |
|

INS
Genomic Map