Assay Details
| Gene Symbol: UBE2Q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082694.1 | NP_001076163.1 | 1 | 1849 | 96 | |
| GenBank mRNA | AY330351.1 | - | 1 | 1849 | 96 | |
More Information
Set Membership: |
|

UBE2Q2
Genomic Map