Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101714.1 | NP_001095184.1 | 6 | 648 | 77 | |
| XM_008258418.2 | XP_008256640.1 | 6 | 859 | 77 | ||
| GenBank mRNA | S61902.1 | - | 5 | 575 | 77 | |
| X54708.1 | - | 648 | 77 | |||
More Information
Set Membership: |
|

TFPI
Genomic Map