Assay Details
| Gene Symbol: KCNJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342854.1 | XP_017198343.1 | 1 | 3435 | 74 | |
| GenBank mRNA | AJ408879.1 | - | 1 | 447 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNJ3| Gene Symbol: KCNJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342854.1 | XP_017198343.1 | 1 | 3435 | 74 | |
| GenBank mRNA | AJ408879.1 | - | 1 | 447 | 74 | |
Set Membership: |
|