Assay Details
| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160286.1 | NP_001153758.1 | 1 - 2 | 79 | 103 | |
| GenBank mRNA | AB469827.1 | - | 1 - 2 | 79 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001160286.1 transcripts.
Set Membership: |
|

NPY
Genomic Map