Assay Details
| Gene Symbol: HAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710331.3 | XP_002710377.1 | 1 | 498 | 83 | |
| XM_008255296.2 | XP_008253518.1 | 1 | 500 | 83 | ||
| GenBank mRNA | AJ291309.1 | - | 1 | 149 | 83 | |
More Information
Set Membership: |
|

HAND1
Genomic Map