Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089317.1 | NP_001082786.1 | 504 | 77 | ||
| GenBank mRNA | EF187821.1 | - | 504 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYBB2| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089317.1 | NP_001082786.1 | 504 | 77 | ||
| GenBank mRNA | EF187821.1 | - | 504 | 77 | ||
Set Membership: |
|