Assay Details
| Gene Symbol: SERPINB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171282.1 | NP_001164753.1 | 4 - 5 | 497 | 81 | |
| XM_008261379.2 | XP_008259601.1 | 5 - 6 | 640 | 81 | ||
| XM_008261380.2 | XP_008259602.1 | 5 - 6 | 680 | 81 | ||
| XM_017343648.1 | XP_017199137.1 | 6 - 7 | 740 | 81 | ||
More Information
Set Membership: |
|

SERPINB12
Genomic Map