Assay Details
| Gene Symbol: SERPINB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171282.1 | NP_001164753.1 | 5 - 6 | 646 | 74 | |
| XM_008261379.2 | XP_008259601.1 | 6 - 7 | 789 | 74 | ||
| XM_008261380.2 | XP_008259602.1 | 6 - 7 | 829 | 74 | ||
| XM_017343648.1 | XP_017199137.1 | 7 - 8 | 889 | 74 | ||
More Information
Set Membership: |
|

SERPINB12
Genomic Map