Assay Details
| Gene Symbol: CKMT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171313.1 | NP_001164784.1 | 3 - 4 | 447 | 98 | |
| XM_008268820.2 | XP_008267042.1 | 4 - 5 | 694 | 98 | ||
| XM_008268821.2 | XP_008267043.1 | 4 - 5 | 705 | 98 | ||
| XM_017347911.1 | XP_017203400.1 | 4 - 5 | 646 | 98 | ||
| XM_017347912.1 | XP_017203401.1 | 4 - 5 | 571 | 98 | ||
More Information
Set Membership: |
|

CKMT1B
Genomic Map