Assay Details
| Gene Symbol: CKMT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171313.1 | NP_001164784.1 | 7 - 8 | 1011 | 86 | |
| XM_008268820.2 | XP_008267042.1 | 8 - 9 | 1258 | 86 | ||
| XM_008268821.2 | XP_008267043.1 | 8 - 9 | 1269 | 86 | ||
| XM_017347911.1 | XP_017203400.1 | 8 - 9 | 1210 | 86 | ||
| XM_017347912.1 | XP_017203401.1 | 8 - 9 | 1135 | 86 | ||
More Information
Set Membership: |
|

CKMT1B
Genomic Map