Assay Details
| Gene Symbol: SLC5A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171357.1 | NP_001164828.1 | 7 - 8 | 666 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC5A4| Gene Symbol: SLC5A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171357.1 | NP_001164828.1 | 7 - 8 | 666 | 61 | |
Set Membership: |
|