Assay Details
| Gene Symbol: TFPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361491.1 | NP_001348420.1 | 621 | 57 | ||
| XM_008251646.2 | XP_008249868.2 | 379 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TFPT| Gene Symbol: TFPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361491.1 | NP_001348420.1 | 621 | 57 | ||
| XM_008251646.2 | XP_008249868.2 | 379 | 57 | |||
Set Membership: |
|