Assay Details
| Gene Symbol: MDFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171407.1 | NP_001164878.1 | 3 - 4 | 495 | 65 | |
| XM_008258188.2 | XP_008256410.1 | 4 - 5 | 1039 | 65 | ||
More Information
Set Membership: |
|

MDFIC
Genomic Map