Assay Details
| Gene Symbol: FGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171488.1 | NP_001164959.1 | 1 - 2 | 176 | 77 | |
| XM_008254887.2 | XP_008253109.1 | 2 - 3 | 496 | 77 | ||
| XM_008254888.2 | XP_008253110.1 | 3 - 4 | 761 | 77 | ||
| XM_008254889.2 | XP_008253111.1 | 2 - 3 | 578 | 77 | ||
More Information
Set Membership: |
|

FGF1
Genomic Map