Assay Details
| Gene Symbol: ELAVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185148.2 | NP_001172077.1 | 654 | 70 | ||
| XM_008249127.2 | XP_008247349.1 | 933 | 70 | |||
| GenBank mRNA | HM037267.1 | - | 655 | 70 | ||
More Information
Set Membership: |
|

ELAVL1
Genomic Map