Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190430.1 | NP_001177359.1 | 1 - 2 | 467 | 94 | |
| GenBank mRNA | J04717.1 | - | 1 - 2 | 467 | 94 | |
More Information
Set Membership: |
|

CYP27A1
Genomic Map