Assay Details
| Gene Symbol: KCNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199008.1 | NP_001185937.1 | 1 | 1750 | 80 | |
| XM_008268561.2 | XP_008266783.1 | 2 | 2870 | 80 | ||
More Information
Set Membership: |
|

KCNA4
Genomic Map