Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204215.1 | NP_001191144.1 | 3 - 4 | 2666 | 69 | |
| NM_001370834.1 | NP_001357763.1 | 9 - 10 | 3911 | 69 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map