Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204216.1 | NP_001191145.1 | 1 - 2 | 210 | 68 | |
| NM_001370834.1 | NP_001357763.1 | 1 - 2 | 210 | 68 | ||
| XM_008259550.2 | XP_008257772.1 | 2 - 3 | 167 | 68 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map