Assay Details
| Gene Symbol: CENPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204933.1 | NP_001191862.1 | 3 - 4 | 210 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPS| Gene Symbol: CENPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204933.1 | NP_001191862.1 | 3 - 4 | 210 | 91 | |
Set Membership: |
|