Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199819.1 | NP_001186748.1 | 1 - 2 | 166 | 56 | |
| GenBank mRNA | DN887612.1 | - | 1 - 2 | 166 | 56 | |
More Information
Set Membership: |
|

SELENOT
Genomic Map