Assay Details
| Gene Symbol: LOC100328967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171270.2 | NP_001164741.2 | 8 | 1218 | 90 | |
| GenBank mRNA | K02441.1 | - | 8 | 1250 | 90 | |
More Information
Set Membership: |
|

LOC100328967
Genomic Map