Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723734.3 | XP_002723780.1 | 567 | 56 | ||
| XM_017339822.1 | XP_017195311.1 | 361 | 56 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMP3| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723734.3 | XP_002723780.1 | 567 | 56 | ||
| XM_017339822.1 | XP_017195311.1 | 361 | 56 | |||
Set Membership: |
|