Assay Details
| Gene Symbol: LOC100342335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273836.2 | XP_008272058.1 | 2640 | 96 | ||
| XM_008273837.2 | XP_008272059.1 | 2582 | 96 | |||
| XM_008273838.2 | XP_008272060.1 | 2624 | 96 | |||
| XM_017350278.1 | XP_017205767.1 | 2993 | 96 | |||
More Information
Set Membership: |
|

LOC100342335
Genomic Map