Assay Details
| Gene Symbol: RGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720984.3 | XP_002721030.1 | 924 | 61 | ||
| XM_017350530.1 | XP_017206019.1 | 946 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720984.3 | XP_002721030.1 | 924 | 61 | ||
| XM_017350530.1 | XP_017206019.1 | 946 | 61 | |||
Set Membership: |
|