Assay Details
| Gene Symbol: GABRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709416.3 | XP_002709462.1 | 3 - 4 | 455 | 83 | |
| XM_017337540.1 | XP_017193029.1 | 3 - 4 | 456 | 83 | ||
| XM_017337541.1 | XP_017193030.1 | 2 - 3 | 84 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017337541.1 transcripts.
Set Membership: |
|

GABRA4
Genomic Map