Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258041.2 | XP_008256263.1 | 1 - 2 | 1015 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008258041.2 transcripts.
Set Membership: |
|

CHRM2
Genomic Map