Assay Details
| Gene Symbol: ALG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720603.3 | XP_002720649.2 | 311 | 66 | ||
| XM_008273561.2 | XP_008271783.1 | 311 | 66 | |||
| XM_017350157.1 | XP_017205646.1 | 311 | 66 | |||
More Information
Set Membership: |
|

ALG5
Genomic Map