Assay Details
| Gene Symbol: CCDC28B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720632.3 | XP_002720678.1 | 340 | 53 | ||
| XM_017350197.1 | XP_017205686.1 | 1166 | 53 | |||
| XM_017350198.1 | XP_017205687.1 | 776 | 53 | |||
| XM_017350199.1 | XP_017205688.1 | 286 | 53 | |||
More Information
Set Membership: |
|

CCDC28B
Genomic Map