Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270431.2 | XP_008268653.1 | 6 - 7 | 1509 | 66 | |
| XM_008270487.2 | XP_008268709.1 | 7 - 8 | 820 | 66 | ||
| XM_008270539.2 | XP_008268761.1 | 7 - 8 | 834 | 66 | ||
| XM_008270600.2 | XP_008268822.1 | 6 - 7 | 676 | 66 | ||
| XM_017340240.1 | XP_017195729.1 | 5 - 6 | 1350 | 66 | ||
More Information
Set Membership: |
|

FAM214B
Genomic Map