Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708810.3 | XP_002708856.1 | 3 - 4 | 1236 | 62 | |
| XM_017345344.1 | XP_017200833.1 | 2 - 3 | 268 | 62 | ||
More Information
Set Membership: |
|

ADM
Genomic Map