Assay Details
| Gene Symbol: RAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711319.3 | XP_002711365.1 | 4 - 5 | 268 | 99 | |
| XM_017341970.1 | XP_017197459.1 | 4 - 5 | 268 | 99 | ||
More Information
Set Membership: |
|

RAP1B
Genomic Map