Assay Details
| Gene Symbol: SERTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350177.1 | XP_017205666.1 | 388 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017350177.1 transcripts.
Set Membership: |
|

SERTM1
Genomic Map