Assay Details
| Gene Symbol: ZBED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250570.2 | XP_008248792.1 | 527 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008250570.2 transcripts.
Set Membership: |
|

ZBED1
Genomic Map