Assay Details
| Gene Symbol: LOC100343059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344663.1 | XP_017200152.1 | 3 - 4 | 773 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100343059| Gene Symbol: LOC100343059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344663.1 | XP_017200152.1 | 3 - 4 | 773 | 78 | |
Set Membership: |
|