Assay Details
| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715054.3 | XP_002715100.2 | 1 - 2 | 1642 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHPF2| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715054.3 | XP_002715100.2 | 1 - 2 | 1642 | 87 | |
Set Membership: |
|