Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262406.2 | XP_008260628.1 | 8 - 9 | 680 | 64 | |
| XM_008262408.2 | XP_008260630.1 | 7 - 8 | 597 | 64 | ||
More Information
Set Membership: |
|

MCUR1
Genomic Map