Assay Details
| Gene Symbol: GPR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712038.3 | XP_002712084.1 | 1 - 2 | 402 | 110 | |
| XM_008258348.2 | XP_008256570.1 | 1 - 2 | 402 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712038.3, XM_008258348.2 transcripts.
Set Membership: |
|

GPR22
Genomic Map