Assay Details
| Gene Symbol: GPR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712038.3 | XP_002712084.1 | 2 - 3 | 1325 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712038.3 transcripts.
Set Membership: |
|

GPR22
Genomic Map